missing translation for 'onlineSavingsMsg'
Learn More
Iodoform, 99%, Thermo Scientific Chemicals Product Code.: 11400264

Iodoform, 99%, Thermo Scientific Chemicals

Product Code. 11400264
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11400264

Brand: Thermo Scientific Alfa Aesar A13664.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

Solubility
Soluble in water, ether, acetic acid, ethanol, chloroform, ether, glycerol, carbon disulfide, olive oil, benzene and acetone. Slightly soluble in petroleum ether.

Notes
Light sensitive. Incompatible with strong bases, alkali metals, trong oxidizing agents and magnesium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-47-8
Molecular Formula CHI3
Molecular Weight (g/mol) 393.732
MDL Number MFCD00001069
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
Synonym triiodomethane, methane, triiodo, carbon triiodide, jodoform, trijodmethane, dezinfekt v, jodoform czech, trijodmethane czech, ccris 346, unii-kxi2j76489
PubChem CID 6374
ChEBI CHEBI:37758
IUPAC Name iodoform
SMILES C(I)(I)I

Specifications

Melting Point 118°C to 123°C
Density 4.008
Flash Point None
Odor Unpleasant
Quantity 50 g
Beilstein 1697010
Sensitivity Light sensitive
Merck Index 14,5033
Solubility Information Soluble in water,ether,acetic acid,ethanol,chloroform,ether,glycerol,carbon disulfide,olive oil,benzene and acetone. Slightly soluble in petroleum ether.
Formula Weight 393.73
Percent Purity 99%
Chemical Name or Material Iodoform
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoform, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.