Learn More
Iodomethane, 2M soln. in tert-butyl methyl ether, Thermo Scientific Chemicals
85.07 € - 248.00 €
Chemical Identifiers
CAS | 74-88-4 |
---|---|
Molecular Formula | CH3I |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00001073 |
InChI Key | INQOMBQAUSQDDS-UHFFFAOYSA-N |
Synonym | methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek |
PubChem CID | 6328 |
ChEBI | CHEBI:39282 |
IUPAC Name | iodomethane |
SMILES | CI |
Description
Iodomethane is used to control insects, plant parasitic nematodes, soil borne pathogens and weed seeds. Iodomethane as a potential metal mobilizing agent in nature. Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodomethane is used to control insects, plant parasitic nematodes, soil borne pathogens and weed seeds. Iodomethane as a potential metal mobilizing agent in nature. Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, bases.
Chemical Identifiers
74-88-4 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CHEBI:39282 | |
CI |
Specifications
74-88-4 | |
41°C to 43°C | |
MFCD00001073 | |
Light sensitive | |
Insoluble in water. | |
CI | |
141.94 | |
CHEBI:39282 | |
2M soln. in tert-butyl methyl ether |
0.933 | |
CH3I | |
UN1992 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
6328 | |
141.94 | |
Iodomethane |
Safety and Handling
GHS H Statement
H225-H301-H331-H351-H315-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P372-P374-P38
H225-H301+H331-H315-H320-H335-H351
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 200-819-5
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only