missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™

Product Code. p-3212763 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10473112

Brand: Thermo Scientific Acros 378411000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 74-88-4 | CH3I | 141.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iodomethane
Name Note 2M Solution in tert-Butyl Ether
CAS 74-88-4, 64-19-7
Density 0.9330g/mL
Boiling Point 41.0°C to 43.0°C
Flash Point −18°C
Packaging AcroSeal™ Glass bottle
Molecular Formula CH3I
MDL Number MFCD00001073
Quantity 100 mL
Merck Index 15, 6159
Specific Gravity 0.933
Synonym mercury 2+ ion acetic acid
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]
IUPAC Name mercury(2+);diacetate
Molecular Weight (g/mol) 141.94
PubChem CID 15337
ChEBI CHEBI:33211
Formula Weight 141.94
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.