missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylsilane, 99%, Thermo Scientific Chemicals

Product Code. 11451960
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4206-67-1
Molecular Formula C4H11ISi
Molecular Weight (g/mol) 214.121
MDL Number MFCD00001077
InChI Key VZNYXGQMDSRJAL-UHFFFAOYSA-N
Synonym iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
PubChem CID 77877
IUPAC Name iodomethyl(trimethyl)silane
SMILES C[Si](C)(C)CI

Specifications

Boiling Point 140°C to 142°C
Quantity 10 g
Linear Formula (CH3)3SiCH2I
UN Number UN1993
Beilstein 1731485
Flash Point 31°C (88°F)
Solubility Information Difficult to mix.
Formula Weight 214.12
Percent Purity 99%
Odor Recognizable
Refractive Index 1.4915
Sensitivity Light sensitive
Density 1.443
Chemical Name or Material (Iodomethyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.