missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Iodonitrotetrazolium violet, 95%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Used for dehydrogenase determination
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∼231°C (decomposition) |
Quantity | 1 g |
Beilstein | 4093224 |
Sensitivity | Light sensitive |
Formula Weight | 505.70 (Anhydrous) |
Percent Purity | 95% |
Chemical Name or Material | Iodonitrotetrazolium violet |
Safety and Handling


Product Identifier
- Iodonitrotetrazolium violet
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Iodonitrotetrazoliumviolet
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction