Learn More
Iodotrimethylplatinum(IV), Thermo Scientific Chemicals
243.00 € - 437.00 €
Chemical Identifiers
CAS | 14364-93-3 |
---|---|
Molecular Formula | C3H9IPt |
Molecular Weight (g/mol) | 367.10 |
MDL Number | MFCD00013702 |
InChI Key | ZCSQPOLLUOLHHF-UHFFFAOYSA-M |
Synonym | iodotrimethylplatinum iv, iodotrimethylplatinum, trimethylplatinum iodide, acmc-1c6hk, trimethylplatinum iv iodide |
PubChem CID | 11824720 |
IUPAC Name | carbanide;iodoplatinum |
SMILES | [CH3-].[CH3-].[CH3-].I[Pt] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393688
|
Thermo Scientific Alfa Aesar
039286.02 |
0.5 g |
243.00 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383688
|
Thermo Scientific Alfa Aesar
039286.03 |
1 g |
437.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14364-93-3 | |
367.10 | |
ZCSQPOLLUOLHHF-UHFFFAOYSA-M | |
11824720 | |
[CH3-].[CH3-].[CH3-].I[Pt] |
C3H9IPt | |
MFCD00013702 | |
iodotrimethylplatinum iv, iodotrimethylplatinum, trimethylplatinum iodide, acmc-1c6hk, trimethylplatinum iv iodide | |
carbanide;iodoplatinum |
Specifications
14364-93-3 | |
Crystalline | |
(CH3)3PtI | |
iodotrimethylplatinum iv, iodotrimethylplatinum, trimethylplatinum iodide, acmc-1c6hk, trimethylplatinum iv iodide | |
ZCSQPOLLUOLHHF-UHFFFAOYSA-M | |
carbanide;iodoplatinum | |
11824720 | |
Odorless | |
Iodotrimethylplatinum(IV) |
Orange to White | |
C3H9IPt | |
MFCD00013702 | |
It is insoluble in water.It is sparingly soluble ether,alcohol,acetone or light petroleum. | |
[CH3-].[CH3-].[CH3-].I[Pt] | |
367.10 | |
367.09 | |
Air & Light Sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only