missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals

44.92 € - 969.83 €

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.1
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10583551
View Documents
Thermo Scientific Acros
187490050
5 mL Glass bottle
44.92 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10733001
View Documents
Thermo Scientific Acros
187490250
25 mL Glass bottle
151.76 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10583741
View Documents
Thermo Scientific Acros
187491000
100 mL Glass bottle
456.27 €
100mL
Please to purchase this item. Need a web account? Register with us today!
13468428
View Documents
Thermo Scientific Acros
187492500
250 mL Glass Bottle
969.83 €
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

16029-98-4
200.1
CSRZQMIRAZTJOY-UHFFFAOYSA-N
85247
C[Si](C)(C)I
C3H9ISi
MFCD00001028
trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
iodo(trimethyl)silane

Specifications

16029-98-4
1.4000g/mL
−31°C
95% min. (Ex I) (Argentometry)
(CH3)3SiI
MFCD00001028
1.4
Solubility in water: reacts
C[Si](C)(C)I
copper
85247
95 to 97%
Iodotrimethylsilane, 95-97%
Colorless to Red
106.0°C
Authentic
C3H9ISi
1.4700 to 1.4800
08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
CSRZQMIRAZTJOY-UHFFFAOYSA-N
iodo(trimethyl)silane
200.1
200.1
Liquid
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 240-171-0

missing translation for 'tsca' : TSCA

Nombre del producto
  • Iodotrimethylsilane, stabilized

Palabra de advertencia
  • Danger

Clasificación
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.

Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

Otros peligros
  • MIXTURE LIST-Contains : odotrimethylsilane

SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only