missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simple Cyanide Reference Standard For Ion Chromatography, H2O/2% KOH, 1000μg/mL:KCN, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
71.68 € - 131.00 €
Specifications
CAS Min % | 0.1, 2.0, 97.9 |
---|---|
CAS Max % | 0.1, 2.0, 97.9 |
Solution Type | Standard Solution |
CAS | 7732-18-5 |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Type | Certified Reference Material |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Solvent or Matrix | 2% KOH |
For Use With (Application) | Ion Chromatography |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10192342
|
SPEX Certiprep
RSCN9-2Y |
125 mL | Triple-leached LDPE bottle | N/A | ||||||
10497292
|
SPEX Certiprep
RSCN9-2X |
500 mL | Triple-leached LDPE bottle |
131.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
CN- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
0.1, 2.0, 97.9 | |
Standard Solution | |
With Certificate of Analysis. | |
Certified Reference Material | |
2% KOH | |
For Use In Matrix Contents Cat. No. EPA Methods 335.2 and 335.3; ASTM Method D2036-19; and Standard Method 4500-CNF. CN− in H2O 1000μg/mL:KCN RSCN9-2X, -2Y EPA Method 335.1 CN− in H2O 1000μg/mL:K3Fe(CN)6 RSCN9C-2X |
0.1, 2.0, 97.9 | |
7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Ion Chromatography | |
Cyanide Standard (Simple) |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1002
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H312-Harmful in contact with skin.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium hydroxide,potassium cyanide
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title