missing translation for 'onlineSavingsMsg'
Learn More

Phosphate-P Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

66.05 € - 129.00 €

Specifications

CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
CAS 7732-18-5
Certificate of Analysis With Certificate of Analysis
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) 10002g/mL
Solvent or Matrix H2O
Chemical Name or Material Phosphate-Phosphorus Standard
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15648235
View Documents Promotion Details
SPEX Certiprep
AS-PO4P9-2Y
125 mL Triple-leached LDPE bottle
66.05 €
125mL
Please to purchase this item. Need a web account? Register with us today!
15638235
View Documents Promotion Details
SPEX Certiprep
AS-PO4P9-2X
500 mL Triple-leached LDPE bottle
129.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
PO4-P1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

0.1, 99.9
7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
10002g/mL
Phosphate-Phosphorus Standard
0.1, 99.9
With Certificate of Analysis
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
H2O
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only