Learn More
SI Analytics™ Ion-selector Indicator Electrode
Copper electrode with plug head
Brand: SI Analytics™ 285216273
This item is not returnable.
View return policy
Description
- pH range 2 to 6
Specifications
Copper Electrode | |
0.0006 to 6,400 mg/L | |
2 to 6 | |
0°C to 80°C |