Learn More
Thermo Scientific™ (IOQ) Qualification Service
Qualification service for New Heratherm Chamber 750 L (including documentation).
Brand:  Thermo Scientific™ IOQPCKE89003711
Product Code. 30049054
  • 1484.00 € / 1 unit
Explore more special offers
Description
Description
  • IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
  • Verifies equipment and any other accessories are assembled and installed.
  • Configuration information for each system component gets recorded. Installation of Equipment is NOT Included.
  • OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IQ/OQ Service