Learn More
Cytiva IPG Buffers
Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.
Brand:  Cytiva 17-6004-40
 View more versions of this product
Product Code. 10014104
  • 76.00 € / 1mL
Explore more special offers
Description
Description
  • Eliminates background staining
  • Use with Immobiline DryStrip gels to improve protein solubility
  • Ensures optimal conditions for IEF separation
  • High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Colorless to Light Yellow
3 to 11 nonlinear
Liquid
1mL
Immobiline DryStrip Gels
IPG Buffer
IEF, 2-D Electrophoresis
2° to 8°C