Learn More
Memmert™ Iq/Oq Document With Device-Specific Works Test Data (1 Temperature Value), For Ich750
Includes temperature distribution survey for one free-selectable temperature value.
Brand: Memmert™ ICH750D00127
This item is not returnable.
View return policy
Description
- IQ/OQ document with works test data for chamber
- 9 measuring points (size 30), 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand), PQ check list as support for validation by customer
Specifications
IQ/OQ Documentation |