Learn More
Grant Instruments™ IQ/OQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
238.50 € - 704.00 €
Products 18
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15271186
View Documents
Grant Instruments™
IQOQ QBH
All QBH Block Heater Models
704.00 €
Each
15211196
View Documents
Grant Instruments™
IQOQ R4
Grant R4 Refrigerated Baths/Circulators This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15221196
View Documents
Grant Instruments™
IQOQ SAP
JBA and SBB Unstirred Baths
561.00 €
Each
15251196
View Documents
Grant Instruments™
IQOQ SUB AQUA PLUS
All SUB Aqua Plus Models This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15211186
View Documents
Grant Instruments™
IQOQ BTD
All Grant BTD Block Heater Models
248.00 €
Each
15261186
View Documents
Grant Instruments™
IQOQ QBD
All QBD Block Heater Models
704.00 €
Each
15231186
View Documents
Grant Instruments™
IQOQ JBN JBA
JBN/JBA Baths
561.00 €
Each
15241196
View Documents
Grant Instruments™
IQOQ SBB AQUA PLUS
All SBB Aqua Plus Models
561.00 €
Each
15281196
View Documents
Grant Instruments™
IQOQ TX150
Grant TX150 Stirred Thermostatic Bath/Circulator
704.00 €
Each
15201206
View Documents
Grant Instruments™
IQOQ XUBA
All Grant XUBA Analogue Ultrasonic Baths
561.00 €
Each
15102625
View Documents
Grant Instruments™
IQOQ CS200G
CS200G Stirred Thermostatic Bath/Circulator This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15182615
View Documents
Grant Instruments™
IQOQ CXG
CXG Stirred Thermostatic Bath/Circulator This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15122615
View Documents
Grant Instruments™
IQOQ T100
Grant T100 Stirred Thermostatic Bath/Circulator
561.00 €
Each
15142615
View Documents
Grant Instruments™
IQOQ TC120
Grant TC120 Stirred Thermostatic Bath/Circulator
704.00 €
Each
15162615
View Documents
Grant Instruments™
IQOQ TXF200
Grant TXF200 Stirred Thermostatic Bath/Circulator
704.00 €
Each
15410484
View Documents
Grant Instruments™
IQOQ CVP-2
Grant CVP-2 PCR Plate Centrifuge
464.28 €
Each
15440484
View Documents
Grant Instruments™
IQOQ PHMP-100
Grant PHMP-100 Thermoshaker for Microplates
238.50 €
Each
15480474
View Documents
Grant Instruments™
IQOQ TS-DW
Grant TS-DW Deep Well Plate Thermoshaker
238.50 €
Each
Description
Description
  • Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
  • Operational Qualification (OQ): instrument is functioning in accordance with its specification
  • Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.