Learn More
Tocris Bioscience™ IRAK3 Degrader 23
IRAK3 PROTAC™ Degrader
Brand:  Tocris Bioscience™ 7983/5
308.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18784454
  • 386.00 € / 5mg
Explore more special offers
Description
Description
IRAK3 Degrader 23 is a potent and selective IRAK3 PROTAC™ Degrader (DC50 = 2 nM; Dmax = 98% in THP1 cells and primary macrophages), requires CRBN and IRAK3 binding for Degrader activity. Also inhibits CDK11, CDK8, TRKC and GSG2 (IC50 values are 16 nM, 36 nM, 100 nM and 110 nM respectively).PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
2-(2,6-Dioxo-3-piperidyl)-5-[4-[2-[1-[4-[4-[cis-4-[(5-isopropylpyrrolo[2,1-f][1,2,4]triazin-4-yl)amino]cyclohexyl]-piperazin-1-yl]-4-oxo-butanoyl]-4-piperidyl]ethyl]piperazin-1-yl]-isoindoline-1,3-dione
5 mg
C47H63N11O6
2712600-00-3
Active Degraders
0.98