missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
70.25 € - 947.72 €
Chemical Identifiers
CAS | 7439-88-5 |
---|---|
Molecular Formula | Ir |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD00011062 |
InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
Synonym | ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge |
PubChem CID | 23924 |
ChEBI | CHEBI:49666 |
IUPAC Name | iridium |
SMILES | [Ir] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450141
|
Thermo Scientific Alfa Aesar
038327.06 |
5 g |
70.25 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364447
|
Thermo Scientific Alfa Aesar
038327.14 |
25 g |
275.37 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374447
|
Thermo Scientific Alfa Aesar
038327.22 |
100 g |
947.72 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-88-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
23924 | |
iridium |
Ir | |
MFCD00011062 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
CHEBI:49666 | |
[Ir] |
Specifications
7439-88-5 | |
MFCD00011062 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
Iridium, 0.5% on activated carbon powder, Reduced, nominally 50% water wet |
Ir | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
[Ir] | |
192.22 | |
CHEBI:49666 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title