Learn More
Iridium, 99.9%, (trace metal basis), powder, Thermo Scientific Chemicals
410.04 € - 6066.18 €
Chemical Identifiers
CAS | 7439-88-5 |
---|---|
Molecular Formula | Ir |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD00011062 |
InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
Synonym | ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge |
PubChem CID | 23924 |
ChEBI | CHEBI:49666 |
IUPAC Name | iridium |
SMILES | [Ir] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10614712
|
Thermo Scientific Acros
198095000 |
500 mg | Glass bottle |
410.04 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713534
|
Thermo Scientific Acros
198090100 |
10 g | Glass bottle |
6066.18 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7439-88-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
23924 | |
iridium |
Ir | |
MFCD00011062 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
CHEBI:49666 | |
[Ir] |
Specifications
Gray | |
99.9% | |
MFCD00011062 | |
15, 5129 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
192.2 | |
Amorphous Powder |
7439-88-5 | |
Ir | |
02,228; 09,257 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
[Ir] | |
192.22 | |
CHEBI:49666 | |
Trace Metal Basis | |
Iridium |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Iridium, powder
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
Hazard Statement
- H319-Causes serious eye irritation.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only