missing translation for 'onlineSavingsMsg'
Learn More
Iridium, AAS standard solution, Specpure™ Ir 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11386349

Iridium, AAS standard solution, Specpure™ Ir 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11386349
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11386349

Brand: Thermo Scientific Alfa Aesar 088072.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011062

Specifications

Quantity 500 mL
UN Number UN1789
Physical Form Liquid
Concentration IrCl3 in 20% HCl
Grade Specpure™
Chemical Name or Material Iridium, AAS standard solution, Ir 1000μg/mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium, AAS standard solution, Specpure™ Ir 1000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Votre commentaire a été soumis