missing translation for 'onlineSavingsMsg'
Learn More

Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11336428
Click to view available options
Quantity:
0.5 g
2 g
10 g

The primary use of iridium is as a hardening agent in platinum alloys. Other uses are for the making of crucibles and devices that require high temperatures. Osmium/iridium alloys are used for compass bearings. Iridium is commonly used in complexes like Ir(mppy)3 and other complexes in polymer LED technology to increase the efficiency from 25% to about 100% due to triplet harvesting. Used in high-dose radiation therapy for the treatment of prostate and other forms of cancer. Iridium is used in tips of ballpoint pens. Iridium is used as a catalyst for carbonylation of methanol to create acetic acid. At one time, iridium as an alloy with platinum, was used in bushing the vents of heavy ordnance and, in a finely powdered condition (iridium black) for painting porcelain black.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Quantity 0.5 g
UN Number UN3089
Solubility Information Soluble in aqua regia.
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Iridium black

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.