missing translation for 'onlineSavingsMsg'
Learn More

Iridium foil, 0.127mm (0.005in) thick, hard, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11311259
Click to view available options
Quantity:
25 x 25 mm

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-88-5
Assay Percent Range 99.8% (metals basis)
Molecular Formula Ir
MDL Number MFCD00011062
Quantity 25 x 25 mm
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES [Ir]
IUPAC Name iridium
Molecular Weight (g/mol) 192.22
PubChem CID 23924
ChEBI CHEBI:49666
Chemical Name or Material Iridium foil
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'