missing translation for 'onlineSavingsMsg'
Learn More
Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11321259

Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11321259
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
This item is not returnable. View return policy

Product Code. 11321259

Brand: Thermo Scientific Alfa Aesar 011427.FF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 3.5g/25 x 25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Assay Percent Range 99.8% (metals basis)
Quantity 25 x 25 mm
Percent Purity 99.8%
Weight ≈3.5g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Iridium foil, 0.25mm (0.01 in.) thick, Hard, Temper: as rolled

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX19I015 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog Number011426.FF,011427.FF,011426.FI
Lot NumberM06I007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011428.FF,011427.FF,011427.FI
Lot NumberI01Q09 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number011426.FF,011427.FF,011426.FI
Lot NumberD28T037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011426.FF,011427.FF,011426.FI
Lot NumberF07Q24 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011426.FF,011427.FF,011426.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium foil, 0.25mm (0.01in) thick, hard, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.