Learn More
Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 15635-87-7 |
---|---|
Molecular Formula | C15H21IrO6 |
Molecular Weight (g/mol) | 489.54 |
MDL Number | MFCD00015353 |
InChI Key | HLYTZTFNIRBLNA-LNTINUHCSA-K |
Synonym | Iridium(III) acetylacetonate |
PubChem CID | 117590592 |
SMILES | [Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Description
It is used as a photoreducing agent, a precursor for preparing Ir-C thin film and a catalyst for hydrogenation of sulfur containing compounds. Used in various catalysis reactions in organic synthesis, including carbon nanostructures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15635-87-7 | |
489.54 | |
HLYTZTFNIRBLNA-LNTINUHCSA-K | |
117590592 |
C15H21IrO6 | |
MFCD00015353 | |
Iridium(III) acetylacetonate | |
[Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
269°C to 271°C | |
Orange-Yellow | |
C15H21IrO6 | |
Iridium(III) acetylacetonate | |
HLYTZTFNIRBLNA-LNTINUHCSA-K | |
iridium(3+) tris((2Z)-4-oxopent-2-en-2-olate) | |
117590592 | |
Ir 37.5% min. | |
Iridium(III) 2,4-pentanedionate |
15635-87-7 | |
Crystalline | |
MFCD00015353 | |
Insoluble in water; soluble in chloroform and benzene; slightly soluble in ethanol and methanol. | |
[Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
489.54 | |
489.53 | |
Odorless |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 239-711-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only