missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
5 g
Description
Iridium(IV) chloride is used in chemical analysis as testing for nitric acid in the presence of nitrous acid, microscopy and in plating solutions. It acts as a catalyst precursor for the stereoselective reduction of cyclohexanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Physical Form | Glassy Amorphous Pieces, Powder |
Quantity | 0.25 g |
Assay Percent Range | 99.95% (Metal basis) |
Solubility Information | Soluble in water,alcohol and dilute hydrochloric acid. |
Formula Weight | 334.01 |
Sensitivity | Hygroscopic |
Appearance | Drak brown to black glassy amorphous pieces/powder |
Chemical Name or Material | Iridium(IV) chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'