Learn More
Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min, Thermo Scientific Chemicals
221.10 €
Chemical Identifiers
CAS | 7790-45-6 |
---|---|
Molecular Formula | I4Ir |
Molecular Weight (g/mol) | 699.84 |
MDL Number | MFCD00049960 |
InChI Key | UXDVYGYWEFPPTQ-UHFFFAOYSA-N |
Synonym | iridium iv iodide, iridium iodide iri4, iridium iv iodide, premion |
PubChem CID | 25113453 |
IUPAC Name | tetraiodoiridium |
SMILES | [I].[I].[I].[I].[Ir] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314028
|
Thermo Scientific Alfa Aesar
040424.03 |
1 g |
221.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324028
|
Thermo Scientific Alfa Aesar
|
5 g |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Methanol carbonylation to acetic acid is catalysed with high rates at low water concentrations using an iridium/iodide based catalyst. Iridium(IV)-iodide mixtures participates in the Investigations in platinum metal group electrochemistry. Rhodium- and iridium-iodide catalysts also promote the carbonylation of higher alcohols to linear or branched chain carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-45-6 | |
699.84 | |
UXDVYGYWEFPPTQ-UHFFFAOYSA-N | |
25113453 | |
[I].[I].[I].[I].[Ir] |
I4Ir | |
MFCD00049960 | |
iridium iv iodide, iridium iodide iri4, iridium iv iodide, premion | |
tetraiodoiridium |
Specifications
7790-45-6 | |
I4Ir | |
iridium iv iodide, iridium iodide iri4, iridium iv iodide, premion | |
UXDVYGYWEFPPTQ-UHFFFAOYSA-N | |
tetraiodoiridium | |
25113453 | |
99.95% | |
Ir 27.0% min. | |
Iridium(IV) iodide |
Powder | |
MFCD00049960 | |
Insoluble in water and alcohol; Soluble in aqueous KI | |
[I].[I].[I].[I].[Ir] | |
699.84 | |
699.82 | |
Premion™ | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 232-206-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.