missing translation for 'onlineSavingsMsg'
Learn More

Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals

Product Code. 11385667
Click to view available options
Quantity:
1 g

Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-49-8
Molecular Formula CH4
Molecular Weight (g/mol) 16.04
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD31654066 MFCD06411993 MFCD07370731
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
IUPAC Name methane
SMILES C

Specifications

Color Black
Physical Form Powder
Quantity 1 g
UN Number UN1479
Solubility Information Insoluble in water.
Formula Weight 224.2
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Ir 84.5% min
Odor Odorless
Density 11.7 g/mL
Assay (metals basis)
Chemical Name or Material Iridium (IV) oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iridium(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Iridium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.