missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7439-88-5 |
---|---|
Molecular Formula | Ir |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD00011062 |
InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
Synonym | ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge |
PubChem CID | 23924 |
ChEBI | CHEBI:49666 |
IUPAC Name | iridium |
SMILES | [Ir] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-88-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
23924 | |
iridium |
Ir | |
MFCD00011062 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
CHEBI:49666 | |
[Ir] |
Specifications
7439-88-5 | |
Powder | |
MFCD00011062 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
99.95% | |
Iridium powder |
Gray | |
Ir | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
[Ir] | |
192.22 | |
CHEBI:49666 | |
(metals basis) |
Safety and Handling
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-095-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only