missing translation for 'onlineSavingsMsg'
Learn More

Iridium sponge, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11386418
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

UN Number UN3089
Quantity 0.5 g
Percent Purity 99.95%
Mesh Size −100 Mesh
Form Sponge
Assay (metals basis)
Chemical Name or Material Iridium sponge

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'