missing translation for 'onlineSavingsMsg'
Learn More

Iridium Standard For ICP/MS, SPEX CertiPrep™

Product Code. 15235936
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ir1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7439-88-5
Packaging Triple-leached LDPE bottle
Quantity 125 mL
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Ir
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES [Ir]
IUPAC Name iridium
Molecular Weight (g/mol) 192.22
PubChem CID 23924
ChEBI CHEBI:49666
Concentration or Composition (by Analyte or Components) 1mg/L
Solvent or Matrix 2% HCl
Chemical Name or Material Iridium Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'