missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium wire, 0.075mm (0.003in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
433.00 €
Chemical Identifiers
CAS | 7439-88-5 |
---|---|
Molecular Formula | Ir |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD00011062 |
InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
Synonym | ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge |
PubChem CID | 23924 |
ChEBI | CHEBI:49666 |
IUPAC Name | iridium |
SMILES | [Ir] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-88-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
23924 | |
iridium |
Ir | |
MFCD00011062 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
CHEBI:49666 | |
[Ir] |
Specifications
7439-88-5 | |
MFCD00011062 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
99.9% | |
Wire | |
Iridium wire, 0.075mm (0.003 in.) dia. |
Ir | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
[Ir] | |
192.22 | |
CHEBI:49666 | |
≈0.099g/m | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 231-095-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only