missing translation for 'onlineSavingsMsg'
Learn More

Iridium wire, 0.25mm (0.01in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11301269
Click to view available options
Quantity:
10 cm
50 cm
200 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Approx.1.11g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Color White to Yellow
Appearance White to pale yellow wire
Assay Percent Range 99.8% (Metals basis)
Quantity 10 cm
Form Wire
Chemical Name or Material Iridium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'