missing translation for 'onlineSavingsMsg'
Learn More
Irinotecan hydrochloride trihydrate, Thermo Scientific Chemicals Product Code.: 15597729

Irinotecan hydrochloride trihydrate, Thermo Scientific Chemicals

Product Code. 15597729
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15597729

Brand: Thermo Scientific Acros 461540010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136572-09-3
Molecular Formula HCl·3H2O
Molecular Weight (g/mol) 677.18
InChI Key KLEAIHJJLUAXIQ-JDRGBKBRSA-N
Synonym irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11Show MoreShow Less
PubChem CID 60837
SMILES CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl

Specifications

Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 677.18
Percent Purity ≥96.0% (HPLC)
Chemical Name or Material Irinotecan hydrochloride trihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461545 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number461540010,461542500
Lot NumberA0427025 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number461540010,461542500
Lot NumberA0373630 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number461540010,461542500
Lot NumberA0419243 Certificate TypeCertificate of Analysis Date19/05/2020 Catalog Number461540010,461542500
compliance-icons
Product Identifier
  • Irinotecan hydrochloride trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Irinotecan hydrochloride trihydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.