missing translation for 'onlineSavingsMsg'
Learn More

Iron foil, 1.0mm (0.04in) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11337087
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.995%
Grade Puratronic™
Weight ≈4.9g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Iron foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'