missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10467010
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Ferric chloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Gray to Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 97.5 to 102.5% (Iodometry)
Linear Formula FeCl3
Fieser 01,392; 02,199; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 10,185; 11,237; 12,1; 13,133; 14,164; 16,167; 17,138
Merck Index 15,4046
Solubility Information Solubility in water: 480g/L (20°C). Other solubilities: readily soluble in alcohol, ether and acetone, slightly soluble in cs2, practically insoluble in ethyl acetate
Formula Weight 162.21
Percent Purity 98%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Iron(III) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • EUH208-Contains Nickel dichloride. May produce an allergic reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.