missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals Product Code.: 11301739

Iron(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11301739
100 g
Click to view available options
Quantity:
10 g
100 g
1 kg
5 kg
25 kg
This item is not returnable. View return policy

Product Code. 11301739

Brand: Thermo Scientific Alfa Aesar 012357.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Ferric chloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 316°C
Melting Point 306°C
Color Black
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98%
Solubility Information Freely soluble in methanol,ethanol,ether,and acetone.
Formula Weight 162.21
Odor Odorless
Sensitivity Hygroscopic
Appearance Black crystalline
Density 2.9 g/cm3
Chemical Name or Material Iron(III) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.