missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals

Product Code. 11353658
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Iron(III) hexacyanoferrate(II) is uses extensively as an Iron Oxide dye. Prussian blue is used as a paint and wallpaper printing, chemical coatings, carbon paper and in the plastics industry. as an antidote for poisoning with radioactive cesium or thallium. In the metalworking and mechanical engineering Prussian blue is thinly applied as a paste on metal surfaces in order to assess the quality scraped surfaces can.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14038-43-8
Molecular Formula C18Fe7N18
Molecular Weight (g/mol) 859.239
MDL Number MFCD00135663
InChI Key DCYOBGZUOMKFPA-UHFFFAOYSA-N
Synonym prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau
PubChem CID 2724251
IUPAC Name iron(2+);iron(3+);octadecacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3]

Specifications

Color Blue
Physical Form Powder
Quantity 25 g
Solubility Information It is practically insoluble in water.
Formula Weight 859.25
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Iron(III) hexacyanoferrate(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'