Learn More
Iron(III) isopropoxide, 98%, Thermo Scientific Chemicals
102.00 € - 1557.00 €
Chemical Identifiers
CAS | 14995-22-3 |
---|---|
Molecular Formula | C9H21FeO3 |
Molecular Weight (g/mol) | 233.109 |
MDL Number | MFCD00070436 |
InChI Key | QUHDSMAREWXWFM-UHFFFAOYSA-N |
Synonym | iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe |
PubChem CID | 11961763 |
IUPAC Name | iron(3+);propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396289
|
Thermo Scientific Alfa Aesar
044873.03 |
1 g |
102.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306299
|
Thermo Scientific Alfa Aesar
044873.06 |
5 g |
397.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316299
|
Thermo Scientific Alfa Aesar
044873.14 |
25 g |
1557.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron(III) isopropoxide is applied in the field of nano-materials and thin films, engineering and environmental field. It is employed in catalysis and synthesis, energy conversion and storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
14995-22-3 | |
233.109 | |
QUHDSMAREWXWFM-UHFFFAOYSA-N | |
11961763 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3] |
C9H21FeO3 | |
MFCD00070436 | |
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe | |
iron(3+);propan-2-olate |
Specifications
14995-22-3 | |
Ampule | |
Fe[OCH(CH3)2]3 | |
UN3181 | |
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe | |
QUHDSMAREWXWFM-UHFFFAOYSA-N | |
iron(3+);propan-2-olate | |
11961763 | |
98% | |
Iron (III) isopropoxide |
Gray | |
C9H21FeO3 | |
MFCD00070436 | |
Moisture sensitive | |
Decomposes in water. | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3] | |
233.109 | |
233.11 | |
Solid Mass |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
- Iron(III) isopropoxide
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only