missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Nitrate Nonahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
Iron(III) Nitrate Nonahydrate, Certified AR for Analysis
Specifications
Specifications
Melting Point | 47°C |
Color | Purple |
Physical Form | Solid |
Quantity | 250 g |
IUPAC Name | iron(3+) nonahydrate trinitrate |
Formula Weight | 404g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Iron(III) nitrate nonahydrate
- Warning
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Iron(III) nitrate nonahydrate, Ferric nitrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'