Learn More
Iron(III) oxalate hexahydrate, Thermo Scientific Chemicals
74.80 € - 220.10 €
Chemical Identifiers
CAS | 166897-40-1 |
---|---|
Molecular Formula | C6H12Fe2O18 |
Molecular Weight (g/mol) | 483.834 |
MDL Number | MFCD00054430 |
InChI Key | FWXIZVVTJVNNRX-UHFFFAOYSA-H |
Synonym | ferric oxalate hexahydrate |
PubChem CID | 131849393 |
IUPAC Name | iron(3+);oxalate;hexahydrate |
SMILES | C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.[Fe+3].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307408
|
Thermo Scientific Alfa Aesar
031116.06 |
5 g | N/A | ||||||
11387398
|
Thermo Scientific Alfa Aesar
031116.14 |
25 g |
220.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron(III) oxalate hexahydrate is used as a catalyst for the reductive and oxidative degradation of iopamidol under UV and visible light irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
166897-40-1 | |
483.834 | |
FWXIZVVTJVNNRX-UHFFFAOYSA-H | |
131849393 | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.[Fe+3].[Fe+3] |
C6H12Fe2O18 | |
MFCD00054430 | |
ferric oxalate hexahydrate | |
iron(3+);oxalate;hexahydrate |
Specifications
166897-40-1 | |
MFCD00054430 | |
ferric oxalate hexahydrate | |
FWXIZVVTJVNNRX-UHFFFAOYSA-H | |
iron(3+);oxalate;hexahydrate | |
131849393 | |
Powder |
C6H12Fe2O18 | |
UN3288 | |
Soluble in water. | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.[Fe+3].[Fe+3] | |
483.834 | |
483.84 (375.55 Anhydrous) | |
Iron(III) oxalate hexahydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 220-951-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Iron(III) oxalate hexahydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P363-Wash contaminated clothing before reuse.
- P321-Specific treatment (see on this label).
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.