Learn More
Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals
Description
NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1565°C |
Color | Red to Brown |
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water |
Formula Weight | 159.69 |
Odor | Odorless |
Appearance | Red-brown powder |
Density | 5.24 |
Chemical Name or Material | Iron(III) oxide, cosmetic, Sold in collaboration with Nanophase Technologies Corporation |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.