missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11337648
Click to view available options
Quantity:
250 g
1 kg
5 kg

Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Gray-White
Physical Form Granules, Powder
Quantity 1 kg
Solubility Information Soluble in waterSoluble in water. Sparingly soluble in alcohol.
IUPAC Name diiron(3+) hydrate trisulfate
Formula Weight 399.87 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Appearance Grayish-white powder or granules
Density 2.0-2.1 g/cm3 at 20°C
Chemical Name or Material Iron(III) sulfate hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, iron(3+) salt (3:2), hydrate, sulphuric acid,ferric sulphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'