missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard Metal Solution 1000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Product Code. 10191710
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color Colorless
pH 1
CAS 10421-48-4
Molecular Formula FeN3O9
MDL Number 11003
Quantity 500 mL
Synonym ferric nitrate, iron trinitrate, nitric acid, iron 3+ salt, unii-n8h8402xob, iron nitrate fe no3 3, iron iii nitrate, anhydrous, hsdb 451, iron 3+ trinitrate, nitric acid, iron 3+ salt 3:1
InChI Key VCJMYUPGQJHHFU-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3]
IUPAC Name iron(3+);trinitrate
Molecular Weight (g/mol) 241.857
PubChem CID 25251
Packaging HDPE plastic bottle
Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Iron solution 1000 ppm, standard metal solution in approx. 1M nitric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid, Ferricnitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.