missing translation for 'onlineSavingsMsg'
Learn More

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3, Thermo Scientific Chemicals

21.11 € - 81.10 €

Specifications

Chemical Name or Material Iron standard solution
Concentration 997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
Name Note 1 mg/ml Fe in 2% HNO3
CAS 7439-89-6, 7697-37-2, 7732-18-5
Packaging Plastic bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10103740
View Documents
Thermo Scientific Acros
196051000
100 mL Plastic bottle
21.11 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10266680
View Documents
Thermo Scientific Acros
196055000
500 mL Plastic bottle
81.10 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Iron standard solution
1 mg/ml Fe in 2% HNO3
Plastic bottle
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
55.85
7.86
997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
7439-89-6, 7697-37-2, 7732-18-5
Colorless
MFCD00010999
[Fe]
55.85
CHEBI:82664
AAS
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Iron standard solution

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only