Learn More
Hach Lange™ Iron trace Cuvette Test
Brand:  Hach Lange™ LCK521
Product Code. 10273824
  • 215.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Order Info
dimRange
Specifications
Specifications
20
2°C to 8°C
Iron Trace Cuvette Test
0.01 to 1mg/L Fe
1.10-Phenanthroline