missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 0.5mm (0.02in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11358478
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy
Questo articolo non è restituibile. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifica

Assay Percent Range 99.99% (metals basis)
Quantity 5 m
Chemical Name or Material Iron wire

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'