missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 1.0mm (0.04in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11328478
Click to view available options
Quantity:
50 cm
250 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Assay Percent Range 99.99% (metals basis)
Quantity 50 cm
Chemical Name or Material Iron wire

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'