missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Hazard Bags

These low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.

Brand:  Micronova™ BL2-24YEIRBB

 View more versions of this product

Product Code. 30090458

This item is not returnable. View return policy

Description

Description

The Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste

  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
Specifications

Specifications

61 L
Five folded liners per double bag
15.3 in.
Yellow
91 cm
50/Cs.
Not Autoclavable
BLSSH-168
0.050 mm
LDPE
Certificate of Conformance and Certificate of Irradiation Processing
39 cm
16 gal.
24 x 36 in.
61 cm
Sterile
Transporting Biomedical Waste (Non-Sharps)
24 in.
36 in.
Hazard Bag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers