missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Irrigation Sleeve
Irrigation sleeves E 336 for the optimum loading of pipettes/MIS instruments in injector units
Brand: Miele Professional™ 3809390
This item is not returnable.
View return policy
Description
- Pipettes up to max. 445 mm length can be held
- Plastic, screw connection
Specifications
Irrigation Sleeve | |
E 336 | |
121 mm | |
1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction