missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISE Reference Solutions, Fisherbrand™
Shop All Fisherbrand Products
Click to view available options
:
For use with: For refillable double junction ISE electrode
Specifications
Specifications
| Chemical Name or Material | ISE Reference Solution |
| Description | ISE reference solution for refillable double junction ISE electrodes. |
Safety and Handling
Product Identifier
- Potassium chloride solution, 4M, 3.3
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : potassium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction