missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ REGLO-CPF Analog Pump Drive
Brand: Ismatec™ ISM1014B
This item is not returnable.
View return policy
Specifications
Analog | |
0.045 to 180 mL/min. | |
REGLO-CPF | |
135 mm | |
Analog Pump Drive | |
115/230V |
115/230 V 50/60 Hz 50 w | |
50/60 Hz | |
135 mm | |
100 mm | |
50 w | |
18 to 1800 rpm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ismatec™ REGLO-CPF Analog Pump Drive > Depth: 135mm
Spot an opportunity for improvement?Share a Content Correction