missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Cleanroom Masks with Ties
GREENER_CHOICE

ISO Air cleanroom masks have long ties for easy donning and are made with the Mass Balance approach from recycled materials.

Brand:  Hopen™ 60.720M

 View more versions of this product

Product Code. 17734789

  • 115.00 € / Pack of 300
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Headloops improve comfort for the user by making it easier to put on
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Ties
Pleated
Class 100 (ISO 5)
Industry
White
Medium
Grade B
>99%
60.720
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers